Learn More
Sino Biological Recombinant Human CHN1 Protein
Supplier:  Sino Biological 14914HNCB100
Encompass_Preferred
A DNA sequence encoding the human CHN1 (NP_001813.1) (Ala2-Phe459) was fused with two additional amino acids (Gly&Pro) at the N-terminus.
Catalog No. 50-161-9355