Learn More
Sino Biological Recombinant Human Centrin 2 Protein (His Tag)
Supplier:  Sino Biological 14983H08H100
Encompass_Preferred
A DNA sequence encoding the human CETN2 (NP_004335.1) (Met1-Tyr172) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-9369