Learn More
Cayman Chemical 14α-hydroxy Paspalinine, Purity: ≥95%, CAS: 151341-77-4, 1 mg
SDP
Catalog No. 502932866
Encompass_Preferred
A fungal metabolite; dietary administration reduces weight gain of H. zea first instar larvae by 91% at 100 ppm
Catalog No. 50-293-2866 Supplier Cayman Chemical Supplier No. 250811 MG
A fungal metabolite; dietary administration reduces weight gain of H. zea first instar larvae by 91% at 100 ppm