Learn More
Cayman Chemical 14a-hydroxy PaspalInIn 5mg
SDP
Catalog No. 502934994
Encompass_Preferred
A fungal metabolite; dietary administration reduces weight gain of H. zea first instar larvae by 91% at 100 ppm
Catalog No. 50-293-4994 Supplier Cayman Chemical Supplier No. 250815 MG
A fungal metabolite; dietary administration reduces weight gain of H. zea first instar larvae by 91% at 100 ppm