Learn More
Fine Science Tools Crile Hemostats, Stainless Steel, Straight, 14 cm
SDP
Catalog No. NC9247203
Encompass
Tip Shape: Straight/ Tips: Serrated/ Tip Width: 1.5mm/ Clamping Length: 32mm/ Alloy / Material: Stainless Steel/ Length: 14cm
Catalog No. NC9247203 Supplier Fine Science Tools Supplier No. 1300414
Tip Shape: Straight/ Tips: Serrated/ Tip Width: 1.5mm/ Clamping Length: 32mm/ Alloy / Material: Stainless Steel/ Length: 14cm