Learn More
SAI Infusion Technologies 14 Gauge, 2.0" Blunt Needles - Non-sterile, 100/pk
SDP
Catalog No. NC0353208
Encompass
14G2.0" Non-Sterile Blunt Needles with plastic hub and stainless steel blunt cannula. These needles are non-sterile, in a bulk case of 100. ETO Exposed sterile blunt needles are available separately. Product is for laboratory animal research only.
Catalog No. NC0353208 Supplier SAI Infusion Technologies Supplier No. B14200 100 BULK
14G2.0" Non-Sterile Blunt Needles with plastic hub and stainless steel blunt cannula. These needles are non-sterile, in a bulk case of 100. ETO Exposed sterile blunt needles are available separately. Product is for laboratory animal research only.