Learn More
SAI Infusion Technologies 14 Gauge 8.5cm Trocar - Tapered Tip, with Stylet
SDP
Catalog No. NC1393795
Encompass
8.5cm Trocar for Rodent Surgery - 14 Gauge, with a Tapered Tip and Stylet. Subcutaneous tunneling needles (trocars) offer a quick and easy method for routing catheters and electrodes under the skin. SAI's TRO-14MS is manufactured from thick-walled medical-grade stainless steel, has a tapered tip and stylet, and is ideal for mouse surgery. Customized tips and lengths are available, please contact us for more information. 14G, 8.5cm. Product is for laboratory animal research only.
Catalog No. NC1393795 Supplier SAI Infusion Technologies Supplier No. TRO14MS
8.5cm Trocar for Rodent Surgery - 14 Gauge, with a Tapered Tip and Stylet. Subcutaneous tunneling needles (trocars) offer a quick and easy method for routing catheters and electrodes under the skin. SAI's TRO-14MS is manufactured from thick-walled medical-grade stainless steel, has a tapered tip and stylet, and is ideal for mouse surgery. Customized tips and lengths are available, please contact us for more information. 14G, 8.5cm. Product is for laboratory animal research only.