Learn More
MSC Cramer Step Stand Stool, Steel, 16 Inch OAW, 350 lb
Catalog No. 5000533088
Encompass
This step stand stool is constructed from steel and designed to provide stable support with a 350 lb load capacity. It features a 16 inch overall width and a spring-mounted caster base for mobility.
  • Constructed from steel for durability
  • 16 inch overall width
  • 16 inch overall depth
  • 350 lb load capacity
  • Step stand design
  • Spring mounted caster base for easy movement
  • Ships unassembled
Catalog No. 50-005-33088 Supplier MSC Supplier No. 36083517
This step stand stool is constructed from steel and designed to provide stable support with a 350 lb load capacity. It features a 16 inch overall width and a spring-mounted caster base for mobility.
  • Constructed from steel for durability
  • 16 inch overall width
  • 16 inch overall depth
  • 350 lb load capacity
  • Step stand design
  • Spring mounted caster base for easy movement
  • Ships unassembled