Learn More
Cayman Chemical 14-methyl Pentadecanoic Acid methyl ester, Purity: ≥98%, CAS: 5129-60-2, 10 mg
SDP
Catalog No. 502932498
Encompass_Preferred
A methylated fatty acid methyl ester; has been found in S. zeai sea sponges as the fatty acyl component of zeamide, A. indica leaf extract, and C. vulgaris and H. pluvialis microalgae; major component of the vancomycin-induced biofilm produced by VRSA; has been used as a standard for the quantification of 14-methyl pentadecanoic acid in various foods by GC-MS
Catalog No. 50-293-2498 Supplier Cayman Chemical Supplier No. 2481310 MG
A methylated fatty acid methyl ester; has been found in S. zeai sea sponges as the fatty acyl component of zeamide, A. indica leaf extract, and C. vulgaris and H. pluvialis microalgae; major component of the vancomycin-induced biofilm produced by VRSA; has been used as a standard for the quantification of 14-methyl pentadecanoic acid in various foods by GC-MS