Learn More
Beckman Coulter 14 mL Sterile + Certified Free Open-Top Thinwall Ultra-Clear Tube, 14 x 95mm - 48Pk
Catalog No. NC2818550
Encompass_Preferred
This round bottom tube is designed for use in Beckman Coulter instruments and rotors. Ultra-Clear tubes are transparent, allowing for easy location of visibly banded or pelleted samples. Compatible Ultracentrifuge Rotor• Ultracentrifuge: SW 40 TiCertified Free*• Exhibits no detectable DNA, DNase, RNase, PCR inhibitors, or endotoxins• Prevents sample degradation and contamination• Improve QC data qualitySterile • Sterilized via ethylene oxide• Pre-sterilized ready-to-use for improved workflow efficiency • Eliminate need for developing sterilization protocol• Packaged in workflow based quantities • Sterilized and packaged compliant with ISO 11135:2014, and ISO 11607:2006Recommended Applications• Microbiology• Gene therapy• Biotherapeutics• Molecular biology• Cell culture• Plant and animal derived products• Algae Certified Free and Beckman standard tubes also available. *Based on sample results below detectable limits.Certificates available here.
Catalog No. NC2818550 Supplier Beckman Coulter Supplier No. C14294
This round bottom tube is designed for use in Beckman Coulter instruments and rotors. Ultra-Clear tubes are transparent, allowing for easy location of visibly banded or pelleted samples. Compatible Ultracentrifuge Rotor• Ultracentrifuge: SW 40 TiCertified Free*• Exhibits no detectable DNA, DNase, RNase, PCR inhibitors, or endotoxins• Prevents sample degradation and contamination• Improve QC data qualitySterile • Sterilized via ethylene oxide• Pre-sterilized ready-to-use for improved workflow efficiency • Eliminate need for developing sterilization protocol• Packaged in workflow based quantities • Sterilized and packaged compliant with ISO 11135:2014, and ISO 11607:2006Recommended Applications• Microbiology• Gene therapy• Biotherapeutics• Molecular biology• Cell culture• Plant and animal derived products• Algae Certified Free and Beckman standard tubes also available. *Based on sample results below detectable limits.Certificates available here.