Learn More
Chemglass Life Sciences Chemglass Tru-Stir 14mm Shaft Adapter With Collar for Stirrer Shaft Couplings
SDP
Catalog No. 502733537
Encompass_Preferred
The Chemglass 14mm shaft adapter with collar is designed to adapt and securely connect 14 mm stirrer shafts to compatible shaft couplings for laboratory stirring applications. The adapter includes a locking collar to help retain the shaft during operation and is supplied as a single unit.
  • Fits 14 mm stirrer shafts.
  • Includes a collar for secure attachment to the coupling.
  • Used with laboratory stirrer shaft couplings.
  • Sold individually (each).
Catalog No. 50-273-3537 Supplier Chemglass Life Sciences Supplier No. CG204467
The Chemglass 14mm shaft adapter with collar is designed to adapt and securely connect 14 mm stirrer shafts to compatible shaft couplings for laboratory stirring applications. The adapter includes a locking collar to help retain the shaft during operation and is supplied as a single unit.
  • Fits 14 mm stirrer shafts.
  • Includes a collar for secure attachment to the coupling.
  • Used with laboratory stirrer shaft couplings.
  • Sold individually (each).