Learn More
Cayman Chemical 14-methyl Palmitic Acid methyl ester, Purity: ≥98%, CAS: 2490-49-5, 10 mg
SDP
Catalog No. 503016025
Encompass_Preferred
A methylated fatty acid methyl ester; has been found in A. indica leaf extract, S. alboflavus TD-1, and as a minor component in biodiesel produced from C. sorokiniana microalgae; volatile compound released by maize that reduces growth of F. verticillioides; has been used as a standard for the quantification of 14-methyl palmitic acid in various foods by GC-MS
Catalog No. 50-301-6025 Supplier Cayman Chemical Supplier No. 2482110 MG
A methylated fatty acid methyl ester; has been found in A. indica leaf extract, S. alboflavus TD-1, and as a minor component in biodiesel produced from C. sorokiniana microalgae; volatile compound released by maize that reduces growth of F. verticillioides; has been used as a standard for the quantification of 14-methyl palmitic acid in various foods by GC-MS