Learn More
Engel USA 14QT TOP-OPENING FRDG-FRZ-WARM
Catalog No. NC1950978
Encompass_Preferred
14 quart portable top-opening 12/24V DC fridge-freezer-warmer with digital controls
Catalog No. NC1950978 Supplier Engel USA Supplier No. MHD13FDM
14 quart portable top-opening 12/24V DC fridge-freezer-warmer with digital controls