Learn More
Chemglass Life Sciences #14 Loosening Ring
SDP
Catalog No. 501531932
Encompass_Preferred
#14 Loosening Ring. Split Polyamide loosening ring to enable inner and outer Rodaviss members to be pulled apart in the event of freezing.
Catalog No. 50-153-1932 Supplier Chemglass Life Sciences Supplier No. CGB18401
#14 Loosening Ring. Split Polyamide loosening ring to enable inner and outer Rodaviss members to be pulled apart in the event of freezing.