Learn More
Cayman Chemical 14S 15R-EET methyl ester 100ug
SDP
Catalog No. 502937334
Encompass_Preferred
A cytochrome P450 metabolite of arachidonic acid; binds to isolated guinea pig monocytes (Ki = 612.5 nM in a radioligand binding assay); induces dilation of precontracted isolated canine epicardial arterioles (EC50 = 4 pM) and denuded porcine subepicardial arterioles (EC50 = 3 pM)
Catalog No. 50-293-7334 Supplier Cayman Chemical Supplier No. 28631100 UG
A cytochrome P450 metabolite of arachidonic acid; binds to isolated guinea pig monocytes (Ki = 612.5 nM in a radioligand binding assay); induces dilation of precontracted isolated canine epicardial arterioles (EC50 = 4 pM) and denuded porcine subepicardial arterioles (EC50 = 3 pM)