Learn More
Spectrum Pharma Quip Specialties Inc 14X14 BLU CREP DURA RAG 250CS
SDP
Catalog No. 507204425
Encompass_Preferred
Spectrum Pharma Quip Strong, medium-duty, absorbent towels are ideal for tough cleaning tasks. Made in a hydroentangling process with wood pulp and polyester. Can be used wet or dry, again and again. Easily cleans up messy spills including oils and solvents. Soft towels will
Catalog No. 50-720-4425 Supplier Spectrum Pharma Quip Specialties Inc Supplier No. CHI 8789
Spectrum Pharma Quip Strong, medium-duty, absorbent towels are ideal for tough cleaning tasks. Made in a hydroentangling process with wood pulp and polyester. Can be used wet or dry, again and again. Easily cleans up messy spills including oils and solvents. Soft towels will