Learn More
Chemglass Life Sciences Graded Seal, Quartz to Borosilicate, 14mm ID X 16mm OD
SDP
Catalog No. CGQ031013
Encompass_Preferred
Graded Seal, Quartz to Borosilicate, 14mm ID X 16mm OD. Graded seals have 3 inches of quartz sealed to 4 inches of borosilicate.
Catalog No. CGQ031013 Supplier Chemglass Life Sciences Supplier No. CGQ031013
Graded Seal, Quartz to Borosilicate, 14mm ID X 16mm OD. Graded seals have 3 inches of quartz sealed to 4 inches of borosilicate.