Learn More
Chemglass Life Sciences Stopcock, Straight Bore, Glass Shell only for PTFE Plug, 3mm & 4mm Bore, 15.2/30 Barrel Size, 10mm OD Sidearms
SDP
Catalog No. CG42605
Encompass_Preferred
Stopcock, Straight Bore, Glass Shell only for PTFE Plug, 3mm & 4mm Bore, 15.2/30 Barrel Size, 10mm OD Sidearms. Glass portion only of CG-424 stopcock. Accepts 1:5 taper PTFE plugs.
Catalog No. CG42605 Supplier Chemglass Life Sciences Supplier No. CG42605
Stopcock, Straight Bore, Glass Shell only for PTFE Plug, 3mm & 4mm Bore, 15.2/30 Barrel Size, 10mm OD Sidearms. Glass portion only of CG-424 stopcock. Accepts 1:5 taper PTFE plugs.