Learn More
Ace Glass, Inc. 15-25mm PTFE coupling
SDP
Catalog No. NC1830801
Encompass_Preferred
PTFE reducing coupling for threaded columns or fittings. Single o-ring seal for leak tight engagement with hand pressure.
Catalog No. NC1830801 Supplier Ace Glass, Inc. Supplier No. 584362
PTFE reducing coupling for threaded columns or fittings. Single o-ring seal for leak tight engagement with hand pressure.