Learn More
Chemglass Life Sciences 15-425 G.P.I. Glass Thread, 16mm OD Tubing, 100mm OAL
SDP
Catalog No. CG19003
Encompass_Preferred
15-425 G.P.I. Glass Thread, 16mm OD Tubing, 100mm OAL. Thread for replacement on apparatus using G.P.I. (Glass Packaging Institute) thread.PLEASE NOTE: GPI THREADS ARE SOLD IN PACKAGES ONLY.
Catalog No. CG19003 Supplier Chemglass Life Sciences Supplier No. CG19003
15-425 G.P.I. Glass Thread, 16mm OD Tubing, 100mm OAL. Thread for replacement on apparatus using G.P.I. (Glass Packaging Institute) thread.PLEASE NOTE: GPI THREADS ARE SOLD IN PACKAGES ONLY.