Learn More
Rockland Immunochemicals 15/50 Tube TrueBlot Magnetic Separator 1 Each
SDP
Catalog No. 501943991
Encompass_Preferred
A magnetic separation unit designed to hold and separate magnetic particles to the side of the tubes up to five 15 mL and three 50 mL tubes. The unit contains five bar magnets embedded in a plastic housing.
Catalog No. 50-194-3991 Supplier Rockland Immunochemicals Supplier No. TMS1550
A magnetic separation unit designed to hold and separate magnetic particles to the side of the tubes up to five 15 mL and three 50 mL tubes. The unit contains five bar magnets embedded in a plastic housing.