Learn More
Electron Microscopy Sciences Upright Freezer Rack 2 x 3
SDP
Catalog No. 501926808
Encompass_Preferred
Stainless steel racks can accommodate various sizes and styles of storage boxes. Rack features high-durability, corrosion-resistant stainless steel material and handles for easy removal. Designed to fit most freezer models and makes. Holds 15 ml and 50 ml Cardboard Cryogenic Boxes and Transformer Cube.
Catalog No. 50-192-6808 Supplier Electron Microscopy Sciences Supplier No. 41000
Stainless steel racks can accommodate various sizes and styles of storage boxes. Rack features high-durability, corrosion-resistant stainless steel material and handles for easy removal. Designed to fit most freezer models and makes. Holds 15 ml and 50 ml Cardboard Cryogenic Boxes and Transformer Cube.