Learn More
Chemglass Life Sciences Sticky Pad, 17-11/16" x 19-5/8" (450 x 500mm)
SDP
Catalog No. NC1084935
Encompass
Sticky Pad, 17-11/16" x 19-5/8" (450 x 500mm), Sticky pad is an ideal alternative to flask clamps for securing vessels to a shaker platform. The mat is designed for use with containers with a flat wide base. It is ideal for containers such as bottles and dishes that cannot be held with traditional flask clamps.
Catalog No. NC1084935 Supplier Chemglass Life Sciences Supplier No. CLS401016
Add to Cart
Add to Cart
Sticky Pad, 17-11/16" x 19-5/8" (450 x 500mm), Sticky pad is an ideal alternative to flask clamps for securing vessels to a shaker platform. The mat is designed for use with containers with a flat wide base. It is ideal for containers such as bottles and dishes that cannot be held with traditional flask clamps.