Learn More
Medchemexpress LLC 15-Azido-pentadecanoic acid | 118162-46-2 | C15H29N3O2 | 10 MM 1 ML
SDP
Catalog No. 5000372174
Encompass_Preferred
15-Azido-pentadecanoic acid is a click chemistry reagent containing an azide group. It is used to identify and characterize post-translationally palmitylated proteins through a simple and robust two-step labeling and detection technique. This reagent can undergo copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) and strain-promoted alkyne-azide cycloaddition (SPAAC) reactions.
  • Contains an azide group for click chemistry applications
  • Identifies and characterizes post-translationally palmitylated proteins
  • Undergoes copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc)
  • Undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) reactions
Catalog No. 50-003-72174 Supplier Medchemexpress LLC Supplier No. HY15165610MM/1ML
15-Azido-pentadecanoic acid is a click chemistry reagent containing an azide group. It is used to identify and characterize post-translationally palmitylated proteins through a simple and robust two-step labeling and detection technique. This reagent can undergo copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) and strain-promoted alkyne-azide cycloaddition (SPAAC) reactions.
  • Contains an azide group for click chemistry applications
  • Identifies and characterizes post-translationally palmitylated proteins
  • Undergoes copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc)
  • Undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) reactions