Learn More
Medchemexpress LLC 15-azido-pentadecanoic acid | 118162-46-2 | MFCD28396912 | 99.9% | 283.41 g/mol | C15H29N3O2 | 10 MG
SDP
Catalog No. 5000286733
Encompass_Preferred
15-Azido-pentadecanoic acid is an azide-functionalized long-chain fatty acid used as a bioorthogonal probe for lipid labeling and conjugation via click chemistry. The molecule has a terminal azide on a 15-carbon backbone, enabling efficient copper-catalyzed azide-alkyne cycloaddition (CuAAC) to attach fluorescent tags, affinity handles, or linkers for biochemical and cell-based studies.
  • High purity (99.9%).
  • Available as solid and as a 10 mM solution in DMSO.
  • Optimized for copper-catalyzed azide-alkyne cycloaddition (CuAAC).
  • Suitable for lipid labeling, probe synthesis, and conjugation workflows.
  • Molecular formula C15H29N3O2; molecular weight 283.41 g/mol.
Catalog No. 50-002-86733 Supplier Medchemexpress LLC Supplier No. HY15165610MG
15-Azido-pentadecanoic acid is an azide-functionalized long-chain fatty acid used as a bioorthogonal probe for lipid labeling and conjugation via click chemistry. The molecule has a terminal azide on a 15-carbon backbone, enabling efficient copper-catalyzed azide-alkyne cycloaddition (CuAAC) to attach fluorescent tags, affinity handles, or linkers for biochemical and cell-based studies.
  • High purity (99.9%).
  • Available as solid and as a 10 mM solution in DMSO.
  • Optimized for copper-catalyzed azide-alkyne cycloaddition (CuAAC).
  • Suitable for lipid labeling, probe synthesis, and conjugation workflows.
  • Molecular formula C15H29N3O2; molecular weight 283.41 g/mol.