Learn More
Thomas Scientific Upright Freezer Drawer Rack, For 15 x 100 Place Hinged Top Boxes, Holds 15 Boxes (5x3), 17-1/4"L x 12-1/8"H x 6-1/2"D
SDP
Catalog No. 50196052
Encompass_Preferred
Racks are for 100-Cell Hinged Top Plastic Boxes. Drawer Racks allow an easy access to fewer microtube boxes at a time thus protecting other samples in the rack from warming up. Constructed of corrosion resistant stainsteel.
Catalog No. 50-196-052 Supplier Thomas Scientific Supplier No. 1148X08
Racks are for 100-Cell Hinged Top Plastic Boxes. Drawer Racks allow an easy access to fewer microtube boxes at a time thus protecting other samples in the rack from warming up. Constructed of corrosion resistant stainsteel.