Learn More
Medchemexpress LLC 15-Crown-5 | 33100-27-5 | 220.26 | 25 G
SDP
Catalog No. 5000449180
Encompass_Preferred
15-Crown-5 is a biochemical reagent used as an efficient phase transfer catalyst and complexing agent in life science research. It can isolate oxonium ion (H7O3)+ salts, particularly from tetrachloroauric acid solutions, and catalyzes the O-alkylation of sodium salts of carboxylic acids in penicillin and cephalosporin series, aiding esterification without acid. Additionally, it facilitates the Williamson synthesis of ethers with hindered alcohols and sodium hydride.
  • Used with lithium aluminum hydride for reduction reactions in hydrogen solvents.
  • Involved in the Horner-Wadsworth-Emmons reaction to prepare stilbenes from aldehydes.
Catalog No. 50-004-49180 Supplier Medchemexpress LLC Supplier No. 25GHYW007451
15-Crown-5 is a biochemical reagent used as an efficient phase transfer catalyst and complexing agent in life science research. It can isolate oxonium ion (H7O3)+ salts, particularly from tetrachloroauric acid solutions, and catalyzes the O-alkylation of sodium salts of carboxylic acids in penicillin and cephalosporin series, aiding esterification without acid. Additionally, it facilitates the Williamson synthesis of ethers with hindered alcohols and sodium hydride.
  • Used with lithium aluminum hydride for reduction reactions in hydrogen solvents.
  • Involved in the Horner-Wadsworth-Emmons reaction to prepare stilbenes from aldehydes.