Learn More
Strem, An Ascensus Company CAS# 33100-27-5. 10g. 15-Crown-5, 98%. MFCD00005110
SDP
Catalog No. 5090109171
Encompass
CAS# 33100-27-5. 10g. 15-Crown-5, 98%. MFCD00005110. Molecular Weight: 220.26. Molecular Formula: C10H20O5. Color/form: pale yellow liq. Strem# 08-0300. http://www.strem.com/catalog/v/08-0300/
Catalog No. 50-901-09171 Supplier Strem, An Ascensus Company Supplier No. 08030010G
CAS# 33100-27-5. 10g. 15-Crown-5, 98%. MFCD00005110. Molecular Weight: 220.26. Molecular Formula: C10H20O5. Color/form: pale yellow liq. Strem# 08-0300. http://www.strem.com/catalog/v/08-0300/