Learn More
Felix Storch Inc ACCUCOLD 15 cubic feet Chest Freezer - 45
SDP
Catalog No. NC2191049
Encompass_Preferred
This 15.5 cu.ft. chest freezer uses static manual defrost cooling to maintain constant temperature, with an externally controlled digital thermostat to ensure easy temperature monitoring. Extra thick 4 ½" thick insulation offers added efficiency and reliability. -45º C capable chest freezer built with solid insulation for laboratory and life science storage applications. Additional features include a solid, easy-grip handle complete with factory installed lock, a sturdy set of casters, and counterbalanced hinges for comfortable loading and unloading. This unit includes a factory-installed probe hole to allow users to install their own monitoring equipment: https://www.accucold.com/product/EL51LT
This item is not returnable. View return policy
Catalog No. NC2191049 Supplier Felix Storch Inc Supplier No. EL51LT
Add to Cart
Add to Cart
Request a Quote
This 15.5 cu.ft. chest freezer uses static manual defrost cooling to maintain constant temperature, with an externally controlled digital thermostat to ensure easy temperature monitoring. Extra thick 4 ½" thick insulation offers added efficiency and reliability. -45º C capable chest freezer built with solid insulation for laboratory and life science storage applications. Additional features include a solid, easy-grip handle complete with factory installed lock, a sturdy set of casters, and counterbalanced hinges for comfortable loading and unloading. This unit includes a factory-installed probe hole to allow users to install their own monitoring equipment: https://www.accucold.com/product/EL51LT