Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 15-deoxy- 12 14-Pros 1mg | 85235-11-6 | 334.45 g/mol | C20H30O4 | 1 MG
SDP

Catalog No. 5000229250
Encompass_Preferred

15-deoxy-Δ12,14-Prostaglandin D2 is a biologically active metabolite of prostaglandin D2 supplied as a high-purity research standard for in vitro pharmacology and biochemical studies.

  • Purity 98.0%.
  • Molecular formula C20H30O4.
  • Molecular weight 334.45 g/mol.
  • Appearance colorless to light yellow (liquid).
  • Supplied as a 1 mg quantity suitable for analytical and biochemical assays.

Catalog No. 50-002-29250 Supplier Medchemexpress LLC Supplier No. HY1160281MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

15-deoxy-Δ12,14-Prostaglandin D2 is a biologically active metabolite of prostaglandin D2 supplied as a high-purity research standard for in vitro pharmacology and biochemical studies.

  • Purity 98.0%.
  • Molecular formula C20H30O4.
  • Molecular weight 334.45 g/mol.
  • Appearance colorless to light yellow (liquid).
  • Supplied as a 1 mg quantity suitable for analytical and biochemical assays.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.