Learn More
Enzo Life Sciences 15-deoxy-Delta12,14-PGJ2 ELISA kit (1x96wells)
SDP
Supplier:  Enzo Life Sciences ADI900023
Encompass_Preferred
First-to-market 15-d-PGJ2 ELISA commercially available for eicosanoid research. The 15-deoxy-Δ12,14-PGJ2 EIA kit is a colorimetric competitive enzyme immunoassay kit with results in 5 hours. Absorbance is read at 405 nm. This kit saves time and is much less time consuming than GC/MS analysis. Ready-to-use liquid color-coded reagents reduce laboratory error. Alternative Name: 15-deoxy-Delta12,14-Prostaglandin J2. Sensitivity: 36.8 pg/ml (range 195 - 200,000 pg/ml). Assay Time: 5 hours. Applications: ELISA, Colorimetric detection. Species Reactivity: Species independent. Contents: GxR IgG Microtiter plate, Conjugate, Antibody, Assay buffer, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution
Catalog No. 50-658-108