Learn More
Enzo Life Sciences 15-deoxy-Delta12,14-PGJ2 polyclonal antibody (100µl)
SDP
Supplier:  Enzo Life Sciences ADI915043100
Encompass_Preferred
Host: Rabbit. Immunogen: 15-deoxy-Δ12,14-PGJ2 conjugated to a protein carrier molecule. Source: Purified from rabbit serum. Species reactivity: Species independent. Applications: ELISA. Purity: Sodium sulfate precipitation purified. Formulation: Liquid. In PBS containing 1.0mg/ml Rabbit IgG and 0.1% sodium azide.
Catalog No. 50-200-7103