Learn More
Justrite™ 12 Gallon Sure-Grip™ EX Compac Corrosives/Acid Steel Safety Cabinet
GREENER_CHOICE
Catalog No. 19822391 Shop All Justrite Safety Group Products
Change view
Click to view available options
Door Style:
Self-close
Catalog No. Door Style
19-822-391 Self-close
1 options
Catalog No. 19-822-391 Supplier Justrite™ Supplier No. 891222
  • $2,503.00
    $1,541.65 / Each of 1
    Save $961.35
    38% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Increase workplace safety and improve cleaniliness with the proper safety and storage equipment. The Justrite™ 12 Gallon Sure-Grip™ EX Compac Corrosives/Acid Steel Safety Cabinet is specially designed for safely storing acids and corrosives in laboratories.
Countertop and Compac Cabinets let you customize your work areas and make the most of available space. Whether adding to existing storage space or making chemicals readily available at workstation locations, these smaller-sized cabinets are the solution when space is at a premium.
Specifications
Load Bearing Capacity 12 gal.
Door Style Self-close
Color Blue