Learn More
Justrite™ 15 Gallon Under Fume Hood Solvent/Flammable Liquid Safety Cabinet With Manual-Closing Doors
GSA_VAGREENER_CHOICE
Catalog No. 19822320 Shop All Justrite Safety Group Products
Change view
Click to view available options
Color:
Light Neutral
Silver
Yellow
Door Style:
Left-hinged, Manual-close
Right-hinged, Manual-close
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Door Style
19-822-320 Silver Left-hinged, Manual-close
19-822-322 Yellow Left-hinged, Manual-close
19-822-323 Silver Right-hinged, Manual-close
19-822-324 Yellow Right-hinged, Manual-close
19-822-325 Light Neutral Left-hinged, Manual-close
19-822-326 Light Neutral Right-hinged, Manual-close
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 19-822-320 Supplier Justrite™ Supplier No. 882414
Add to Cart
Add to Cart
Increase workplace safety and improve cleaniliness with the proper safety and storage equipment. The Justrite™ 15 Gallon Under Fume Hood Solvent/Flammable Liquid Safety Cabinet With Manual-Closing Doors is fully compliant and designed for safe storage of solvents and flammables in laboratories.
  • Hands-free solvent collection: Solvent flows freely from your process into the container through tubing making it suitable for either pump-fed or gravity-flow HPLC waste methods
  • Two disconnect fittings, one intake and one vent, eliminate back pressure in liquid flow disposal line and provide controlled handling of vapor
  • Durability and safety: Translucent polyethylene container allows quick visual inspection of liquid level
  • Pouring spout is equipped with a stainless steel flame arrester, which reliably dissipates heat to prevent flashback ignition
  • Spring-loaded, sealed lid automatically vents to guard against explosion
  • Control odors with a coalescing/carbon filter
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Load Bearing Capacity 15 gal.
Door Style Left-hinged, Manual-close
Color Silver