Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Justrite™ 15 Gallon Yellow Biohazard Linen Plastic Bags

Catalog No. 17988500 Shop All Justrite Safety Group Products
Change view
Click to view available options
Capacity (English):
15 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English)
17-988-500 15 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-988-500 Supplier Justrite™ Supplier No. 05902

missing translation for 'validMainframeMsg'

Yellow biohazard linen plastic bags minimize waste by segregating infectious linen to be laundered

  • Yellow color easily identifiable as infectious linen bags
  • Printing includes English, Spanish and French translation along with international recognizable biohazard symbol
  • 15 gal. capacity bag fits Justrite™ biohazard cans (6 and 10 gal.)
  • Material: HDPE
  • Thickness: 11 microns
  • Provides secure handling to avoid punctures and tears
  • Tubular construction with single side seal and star seal bottom provide stable, leak proof bag
  • 100/Pk.

Specifications

Capacity (English) 15 gal.
Color Yellow
Autoclavable Not Autoclavable
For Use With (Application) Infectious Linen
Width (English) 33 in.
Length (English) 33 in.
Thickness 11 microns
Height (English) 24 in.
Material HDPE
Product Type Biohazard Linen Bags
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.