Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 15-hexadecynoic acid | 99208-90-9 | MFCD22124928 | 252.39 g/mol | C16H28O2 | 5 MG
SDP

Catalog No. 5000209336
Encompass_Preferred

Alkynyl palmitic acid (15-hexadecynoic acid) is an ω-alkynylated long-chain fatty acid bearing a terminal alkyne. It is used as a click-chemistry reagent and an alkyl-chain PROTAC linker for bio-conjugation and synthesis workflows.

  • Chemical formula: C16H28O2.
  • Molecular weight: 252.39 g/mol.
  • Cas number: 99208-90-9.
  • Contains a terminal alkyne for copper-catalyzed azide-alkyne cycloaddition.
  • Typical applications include PROTAC linker synthesis and bioconjugation.
  • Available pack size: 5 mg.
  • Purity: not specified on the manufacturer datasheet.

Catalog No. 50-002-09336 Supplier Medchemexpress LLC Supplier No. HYW0403045MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Alkynyl palmitic acid (15-hexadecynoic acid) is an ω-alkynylated long-chain fatty acid bearing a terminal alkyne. It is used as a click-chemistry reagent and an alkyl-chain PROTAC linker for bio-conjugation and synthesis workflows.

  • Chemical formula: C16H28O2.
  • Molecular weight: 252.39 g/mol.
  • Cas number: 99208-90-9.
  • Contains a terminal alkyne for copper-catalyzed azide-alkyne cycloaddition.
  • Typical applications include PROTAC linker synthesis and bioconjugation.
  • Available pack size: 5 mg.
  • Purity: not specified on the manufacturer datasheet.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.