Learn More
Chemglass Life Sciences #15 Internal Thread, Glass Only
SDP
Catalog No. 501531987
Encompass_Preferred
#15 Internal Thread, Glass Only. 22mm Tube OD, Accepts 12.5-14mm OD Tubing. Internal glass thread for use with CG-362, CG-363, CG-364, and CG-366 PTFE bushings. Tubing OD listed is the shank OD of the thread.
Catalog No. 50-153-1987 Supplier Chemglass Life Sciences Supplier No. CGB36003
#15 Internal Thread, Glass Only. 22mm Tube OD, Accepts 12.5-14mm OD Tubing. Internal glass thread for use with CG-362, CG-363, CG-364, and CG-366 PTFE bushings. Tubing OD listed is the shank OD of the thread.