Learn More
Medchemexpress LLC HY-W015422 25mg Medchemexpress, 1,5-Isoquinolinediol CAS:5154-02-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW01542225MG
Encompass_Preferred
Medchemexpress, HY-W015422 25mg 1,5-Isoquinolinediol CAS:5154-02-9 1,5-Isoquinolinediol is a potent PARP inhibitor, with an IC50 of 0.18-0.37 µM. 1,5-Isoquinolinediol attenuates diabetes-induced NADPH oxidase-derived oxidative stress in retina. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3376