Learn More
Cayman Chemical 15-keto Fluprostenol 10mg
SDP
Catalog No. 502967441
Encompass_Preferred
A derivative of fluprostenol; a potential inactive metabolite of 15-keto fluprostenol isopropyl ester; a potential metabolite of fluprostenol (travoprost); a potential impurity found in commercial preparations of fluprostenol
Catalog No. 50-296-7441 Supplier Cayman Chemical Supplier No. 1678510 MG
A derivative of fluprostenol; a potential inactive metabolite of 15-keto fluprostenol isopropyl ester; a potential metabolite of fluprostenol (travoprost); a potential impurity found in commercial preparations of fluprostenol