Learn More
Cayman Chemical 15-keto Latnoprost 10mg
SDP
Catalog No. 502965874
Encompass_Preferred
An active metabolite of latanoprost; intraocular administration reduces intraocular pressure in cynomolgus monkeys at 1 UG/eye; reduces pupillary diameter in cats when administered topically at 5 UG/eye
Catalog No. 50-296-5874 Supplier Cayman Chemical Supplier No. 1681610 MG
An active metabolite of latanoprost; intraocular administration reduces intraocular pressure in cynomolgus monkeys at 1 UG/eye; reduces pupillary diameter in cats when administered topically at 5 UG/eye