Learn More
Abcam 15% Lauryl Maltoside Solution, 1ML
Catalog No. 502708147
Encompass_Preferred
n-dodecyl-ß-D-maltopyranoside (lauryl maltoside) solution ab109858 for use with immunocapture, blue native, and dipstick protocols.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-8147 Supplier Abcam Supplier No. AB1098581ML
n-dodecyl-ß-D-maltopyranoside (lauryl maltoside) solution ab109858 for use with immunocapture, blue native, and dipstick protocols.
The product is subject to the following: Abcam Restricted Use Statement