Learn More
Ace Glass, Inc. Beaker, 15L, heavy wall, 260mm OD x 390mm height, not graduated, Duran;
SDP
Catalog No. NC9988979
Encompass
15L Duran heavy wall beaker with spout. Beaker is not graduated. Dimensions: 260mm O.D. x 390mm Height.
Catalog No. NC9988979 Supplier Ace Glass, Inc. Supplier No. 533236
Add to Cart
Add to Cart
15L Duran heavy wall beaker with spout. Beaker is not graduated. Dimensions: 260mm O.D. x 390mm Height.