Learn More
Chemglass Life Sciences Capacity Liters: 15, Flange I.D. (mm): 200, Valve Style: 0-20 Sealed, Overall Height mm (in.): 565 (22.2), Height to Width Ratio: 1.8:1, Inlet/Outlet Size: 1" BP, Jacket Capacity Liters: 7
SDP
Catalog No. CG193032
Encompass_Preferred
Process reaction vessels, jacketed, Capacity Liters: 15, Flange I.D. (mm): 200, Valve Style: 0-20 Sealed, Overall Height mm (in.): 565 (22.2), Height to Width Ratio: 1.8:1, Inlet/Outlet Size: 1" BP, Jacket Capacity Liters: 7. Jacketed large scale process reaction vessels. The 10, 15, & 20L vessels have a 200mm ID flange with an o-ring groove formed in the surface of the flange. The inlet/outlet of the vessels are beaded pipe fittings for use with CG-1964 and CG-1969-A beaded pipe adapters. The 10, 15 and 20L are available with a choice of a 2¿ beaded pipe valve that is detachable or the 0-20mm valve with 3/4¿ sidearm that is permanently sealed. These flush bottom (zero dead space) valves are designed to prevent solids from collecting in the bottom of the vessel. Vessels equipped with the 0-20mm sealed on valve are supplied complete with a TFE plug having Perfluoro chemically resistant o-rings.PLEASE NOTE: VALVE, BEADED PIPE COUPLINGS, AND ADAPTERS MUST BE PURCHASED SEPARATELY.
Catalog No. CG193032 Supplier Chemglass Life Sciences Supplier No. CG193032
Add to Cart
Add to Cart
Request a Quote
Process reaction vessels, jacketed, Capacity Liters: 15, Flange I.D. (mm): 200, Valve Style: 0-20 Sealed, Overall Height mm (in.): 565 (22.2), Height to Width Ratio: 1.8:1, Inlet/Outlet Size: 1" BP, Jacket Capacity Liters: 7. Jacketed large scale process reaction vessels. The 10, 15, & 20L vessels have a 200mm ID flange with an o-ring groove formed in the surface of the flange. The inlet/outlet of the vessels are beaded pipe fittings for use with CG-1964 and CG-1969-A beaded pipe adapters. The 10, 15 and 20L are available with a choice of a 2¿ beaded pipe valve that is detachable or the 0-20mm valve with 3/4¿ sidearm that is permanently sealed. These flush bottom (zero dead space) valves are designed to prevent solids from collecting in the bottom of the vessel. Vessels equipped with the 0-20mm sealed on valve are supplied complete with a TFE plug having Perfluoro chemically resistant o-rings.PLEASE NOTE: VALVE, BEADED PIPE COUPLINGS, AND ADAPTERS MUST BE PURCHASED SEPARATELY.