Learn More
Medchemexpress LLC 15-LOX-1 inhibitor 1 | 2349374-37-2 | 98.8% | 412.87 | 1 MG
SDP
Catalog No. 5000351980
Encompass_Preferred
15-LOX-1 inhibitor 1 is a potent inhibitor of 15-LOX-1 (15-lipoxygenase-1) with an IC50 value of 0.19 μM. It protects macrophages from lipopolysaccharide-induced cytotoxicity and inhibits NO formation and lipid peroxidation. For research use only.
  • Potent inhibitor of 15-LOX-1 (15-lipoxygenase-1)
  • Protects macrophages from lipopolysaccharide-induced cytotoxicity
  • Inhibits NO formation
  • Inhibits lipid peroxidation
  • For research use only
Catalog No. 50-003-51980 Supplier Medchemexpress LLC Supplier No. HY1389891MG
Add to Cart
Add to Cart
15-LOX-1 inhibitor 1 is a potent inhibitor of 15-LOX-1 (15-lipoxygenase-1) with an IC50 value of 0.19 μM. It protects macrophages from lipopolysaccharide-induced cytotoxicity and inhibits NO formation and lipid peroxidation. For research use only.
  • Potent inhibitor of 15-LOX-1 (15-lipoxygenase-1)
  • Protects macrophages from lipopolysaccharide-induced cytotoxicity
  • Inhibits NO formation
  • Inhibits lipid peroxidation
  • For research use only