Learn More
Thermo Scientific 15 MCL INJECTION NEEDLE FOR WP
Catalog No. NC9421732 Shop All Thermo Scientific Products
Encompass
Fused Silica Needle, 15 uL, 250 um i.d. incl. nanoViper Fittings
Catalog No. NC9421732 Supplier Thermo Scientific Supplier No. 6820.3115
Fused Silica Needle, 15 uL, 250 um i.d. incl. nanoViper Fittings