Learn More
Cayman Chemical 15-methyl PalmItIc AcId 10mg
SDP
Catalog No. 502932499
Encompass_Preferred
A methylated fatty acid; has been found in bacteria, bovine milk fat, one-humped camel meat and fat, and in rabbit perirenal lipids; found in human breast milk and levels are decreased in mature breast milk compared to colostrum
Catalog No. 50-293-2499 Supplier Cayman Chemical Supplier No. 2481410 MG
A methylated fatty acid; has been found in bacteria, bovine milk fat, one-humped camel meat and fat, and in rabbit perirenal lipids; found in human breast milk and levels are decreased in mature breast milk compared to colostrum