Learn More
Cayman Chemical 15-methyl Palmitic Acid, Purity: ≥98%, CAS: 1603-03-8, 20 mg
SDP
Catalog No. 502932608
Encompass_Preferred
A methylated fatty acid; has been found in bacteria, bovine milk fat, one-humped camel meat and fat, and in rabbit perirenal lipids; found in human breast milk and levels are decreased in mature breast milk compared to colostrum
Catalog No. 50-293-2608 Supplier Cayman Chemical Supplier No. 2481420 MG
A methylated fatty acid; has been found in bacteria, bovine milk fat, one-humped camel meat and fat, and in rabbit perirenal lipids; found in human breast milk and levels are decreased in mature breast milk compared to colostrum