Learn More
Eurofins Discoverx PathHunter Detection Kit, 15 (2 plates), 200/800 dp (96-well/384-well). (93-0001M)
Catalog No. NC1417417
Encompass
The PathHunter Detection Kit is a detection kit intended for use with Eurofins DiscoverX's PathHunter cell lines or with custom cell lines made using ProLabel/PK expression vectors. This kit contains lysis buffer and EFC substrate to allow for detection of EFC levels in the assayed cells. This kit contains all components required to assay 2 plates (96/384 well). For detailed product information, User manual and COA, visit www.discoverx.com/products/reagents/detection-kit-93-0001m.
Catalog No. NC1417417 Supplier Eurofins Discoverx Supplier No. 930001M
The PathHunter Detection Kit is a detection kit intended for use with Eurofins DiscoverX's PathHunter cell lines or with custom cell lines made using ProLabel/PK expression vectors. This kit contains lysis buffer and EFC substrate to allow for detection of EFC levels in the assayed cells. This kit contains all components required to assay 2 plates (96/384 well). For detailed product information, User manual and COA, visit www.discoverx.com/products/reagents/detection-kit-93-0001m.