Learn More
Andwin Scientific 15 ML CENTRIFUGE TUBE
SDP
Catalog No. NC9634559
Encompass_Preferred
SuperClear Centrifuge Tubes, Polypropylene, 15mL ultraclear tube. Nonsterile. Printed graduations. Printed writing area. Autoclavable. Freezable. Length: 118mm Diameter: 17mm Polypropylene autoclavable cap. Max. RCF: 6,000g.
Catalog No. NC9634559 Supplier Andwin Scientific Supplier No. 21008089
SuperClear Centrifuge Tubes, Polypropylene, 15mL ultraclear tube. Nonsterile. Printed graduations. Printed writing area. Autoclavable. Freezable. Length: 118mm Diameter: 17mm Polypropylene autoclavable cap. Max. RCF: 6,000g.