Learn More
Andwin Scientific 15 ML CENTRIFUGE TUBE WITH CAP
SDP
Catalog No. 50819796
Encompass_Preferred
The 15 mL conical screw cap centrifuge tubes are made with copolymer polypropylene. These tubes feature a large white labeling area that permits easy sample identification and easy-to-read graduations at 1 mL that allows precise measurements. This product comes with high density polyethylene (HDPE) caps that have a sealing ring for a leak-proof seal. It comes in a pack of 500 tubes.
Catalog No. 50-819-796 Supplier Andwin Scientific Supplier No. 14751611
The 15 mL conical screw cap centrifuge tubes are made with copolymer polypropylene. These tubes feature a large white labeling area that permits easy sample identification and easy-to-read graduations at 1 mL that allows precise measurements. This product comes with high density polyethylene (HDPE) caps that have a sealing ring for a leak-proof seal. It comes in a pack of 500 tubes.